Orders LinkContactUsLinkComponent
Test kits, COD, TNT plus®
Test kits, COD, TNT plus®
03, COD reagents, HR, 20 - 1500 mg/l, Reactor digestion, 150
  HACHTNT82206
Test kits, COD, TNT plus®
  HACHTNT82206
 :  
 :  
Test kits, COD, TNT plus®
Test kits, COD, TNT plus®
03, COD reagents, HR, 20 - 1500 mg/l, Reactor digestion, 150
 :  
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process.
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are colour coded by parameter and range
  • No reagent blank necessary
  • 化学方法:
    Reactor digestion
  • 包装规格:
    150
  • 说明:
    COD reagents, HR
  • 顺序:
    03
  • 范围:
    20 - 1500 mg/l