Test kits, COD, TNT plus®
02, COD reagents, LR, 3 - 150 mg/l, Reactor digestion, 25 Tests
HACHTNT821
: HACH
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process.
- Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
- Barcode helps the instrument to automatically detect and run the correct method
- Barcode allows spectrophotometer to detect reagent lot number and expiry date
- Vials have flat bottom to improve handling
- Vials are colour coded by parameter and range
- No reagent blank necessary
- 化学方法:Reactor digestion
- 包装规格:25 Tests
- 说明:COD reagents, LR
- 顺序:02
- 范围:3 - 150 mg/l
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.