Orders LinkContactUsLinkComponent
Test kits, COD, TNT plus®
02, COD reagents, LR, 3 - 150 mg/l, Reactor digestion, 25 Tests
  HACHTNT821
 :   HACH
Test kits, COD, TNT plus®
  HACHTNT821
 :  HACH
 :  
Test kits, COD, TNT plus®
Test kits, COD, TNT plus®
02, COD reagents, LR, 3 - 150 mg/l, Reactor digestion, 25 Tests
 :  
1 * 25 Tests
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process.
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are colour coded by parameter and range
  • No reagent blank necessary
  • 化学方法:
    Reactor digestion
  • 包装规格:
    25 Tests
  • 说明:
    COD reagents, LR
  • 顺序:
    02
  • 范围:
    3 - 150 mg/l