Test kits, COD, TNT plus®
01, COD reagents, ULR, 1 - 60 mg/l, Reactor digestion, 24
Catalog # HACHTNT820
Jump to:
Discontinued
Product Details & Documents
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process.
- Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
- Barcode helps the instrument to automatically detect and run the correct method
- Barcode allows spectrophotometer to detect reagent lot number and expiry date
- Vials have flat bottom to improve handling
- Vials are colour coded by parameter and range
- No reagent blank necessary
Specifications
Specifications
- 化学方法:Reactor digestion
- 包装规格:24
- 说明:COD reagents, ULR
- 顺序:01
- 范围:1 - 60 mg/l