China
Orders LinkContactUsLinkComponent
Test kits, COD, TNT plus®
Test kits, COD, TNT plus®
01, COD reagents, ULR, 1 - 60 mg/l, Reactor digestion, 24
Catalog # HACHTNT820
Test kits, COD, TNT plus®
Catalog # HACHTNT820
Supplier:  
CAS Number:  
Test kits, COD, TNT plus®
Test kits, COD, TNT plus®
01, COD reagents, ULR, 1 - 60 mg/l, Reactor digestion, 24
CAS Number:  
Discontinued
Product Details & Documents
The TNTplus® COD vials are designed to measure COD values in an efficient and accurate process.
  • Dosi-Cap Zip with freeze dried reagents integrated into the sealed cap
  • Barcode helps the instrument to automatically detect and run the correct method
  • Barcode allows spectrophotometer to detect reagent lot number and expiry date
  • Vials have flat bottom to improve handling
  • Vials are colour coded by parameter and range
  • No reagent blank necessary
Specifications
Specifications
  • 化学方法:
    Reactor digestion
  • 包装规格:
    24
  • 说明:
    COD reagents, ULR
  • 顺序:
    01
  • 范围:
    1 - 60 mg/l
Specifications