Orders LinkContactUsLinkComponent
Sampler, StickProof
Sampler, StickProof
Sampler, 1100 mm, 25 mm, ≤50 ml
Catalog # BURK5338-1100
Supplier:   BURKLE
Sampler, StickProof
Catalog # BURK5338-1100
Supplier:  BURKLE
Sampler, StickProof
Sampler, StickProof
Sampler, 1100 mm, 25 mm, ≤50 ml
Price
Inquire For Price
Log in for availability
1 * 1 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Stainless steel sampler for powders. The conical inlet allows for collection of different sample sizes, up to approximately 50 ml. Constructed without edges, grooves or crevices for easy cleaning. The sample is collected directly into the sampling bag which is held in place with the included clamp. StickProof borehole diameter 25 mm; chamber length 120 mm. Sampling bags: 170×120 mm for 400 ml.
  • Especially slender tip can be inserted easily and deeply
  • Electropolished surface
  • Variable sample size up to 50 ml
  • Stainless steel V4A (1.4404)/AISI 316 L
Delivery information: Complete with clamp and 100 sampling bags PE.
Specifications
  • 直径:
    25 mm
  • 容积:
    ≤50 ml
  • 说明:
    Sampler
  • 长度:
    1100 mm
Accessories
Product Information
  • Catalog #
    顺序
    容量
    厚度
    Availability
    Price
  • BURK2348-0003
    03
    140 ml
    0,05
    Log in for availability
    1 * 100 件
Product Information
  • Catalog #
    说明
    Availability
    Price
  • BURK2348-0005
    Packaging bags with closure (120×170 mm), 400 ml
    Log in for availability
    1 * 100 件
Product Information
  • Catalog #
    顺序
    容量
    厚度
    Availability
    Price
  • BURK2348-0010
    09
    6500 ml
    0,05
    Log in for availability
    1 * 100 件
Product Information
  • Catalog #
    顺序
    容量
    厚度
    Availability
    Price
  • BURK2348-0008
    07
    2500 ml
    0,05
    Log in for availability
    1 * 100 件
Product Information
  • Catalog #
    顺序
    容量
    厚度
    Availability
    Price
  • BURK2348-0006
    05
    1000 ml
    0,05
    Log in for availability
    1 * 100 件
Product Information
  • Catalog #
    顺序
    容量
    厚度
    Availability
    Price
  • BURK2348-0009
    08
    4500 ml
    0,05
    Log in for availability
    1 * 100 件
Product Information
  • Catalog #
    顺序
    容量
    厚度
    Availability
    Price
  • BURK2348-0002
    02
    80 ml
    0,05
    Log in for availability
    1 * 100 件
Product Information
  • Catalog #
    顺序
    容量
    厚度
    Availability
    Price
  • BURK2348-0007
    06
    1500 ml
    0,05
    Log in for availability
    1 * 100 件
Product Information
  • Catalog #
    顺序
    容量
    厚度
    Availability
    Price
  • BURK2348-0004
    04
    300 ml
    0,05
    Log in for availability
    1 * 100 件
Product Information
  • Catalog #
    顺序
    容量
    厚度
    Availability
    Price
  • BURK2348-0001
    01
    100 ml
    0,05
    Log in for availability
    1 * 100 件