Conductivity standards
03, Conductivity standard, 12,88 mS/cm, 250 mL
供应商: METTLER TOLEDO
Jump to:
Product Details & Documents
A slight change in temperature usually has a big impact on the conductivity value of a standard solution. A table on every bottle label indicates the conductivity values at the most common measurement temperatures.
During calibration the meter automatically references to this table for temperature compensation. If possible, calibration and measurements should be performed at the same temperature
Documents
规格
规格
- 包装规格:250 mL
- 说明:Conductivity standard
- 顺序:03
- 电导率:12,88 mS/cm