China
Orders LinkContactUsLinkComponent
Conductivity standards
03, Conductivity standard, 12,88 mS/cm, 250 mL
目录编号 METT51350094
供应商:   METTLER TOLEDO
Conductivity standards
目录编号 METT51350094
供应商:  METTLER TOLEDO
CAS号:  
Conductivity standards
Conductivity standards
03, Conductivity standard, 12,88 mS/cm, 250 mL
Jump to:
CAS号:  
Price
询价
Log in 查看库存
1 * 250 毫升
Product Details & Documents
A slight change in temperature usually has a big impact on the conductivity value of a standard solution. A table on every bottle label indicates the conductivity values at the most common measurement temperatures.
During calibration the meter automatically references to this table for temperature compensation. If possible, calibration and measurements should be performed at the same temperature
Documents
Certificates
按批号搜索
规格
规格
  • 包装规格:
    250 mL
  • 说明:
    Conductivity standard
  • 顺序:
    03
  • 电导率:
    12,88 mS/cm
规格