Microplate Lids
Product Details & Documents
Protect your samples with Eppendorf® Microplate Lids, perfect for incubation and short-term storage. Stable, flexible protection available in 'PCR Clean' and 'Sterile' grades, ensuring your samples stay pure and uncontaminated.
- Stable, flexible protection of samples during incubation and short-term storage
规格
- 无菌:非无菌
- 说明:Microplate lid for MTP/DWP, PCR Clean (non sterile), (5 bags×16)
- 顺序:01
- Cat. No.:EPPE0030131.517
- 环境认证:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Low Manufacturing Impact