鞋套,CPE
03, 蓝色, U*
Basic shoe covers manufactured from a low-density chlorinated polyethylene fabric, which is designed to protect against dirt, grime, and liquid splash.
- CPE is water resistant and provides good resistance against chemicals
- Economical and lint-free
Ideal for laboratory use.
* 通用尺寸
- 滑轨尺寸:U*
- 颜色:蓝色
- 顺序:03
- Cat. No.:VWRI113-1492
- Garment application:Laboratory