鞋套,CPE
04, 蓝色, U*
目录编号 VWRU414004-512
Jump to:
Product Details & Documents
Basic shoe covers manufactured from a low-density chlorinated polyethylene fabric, which is designed to protect against dirt, grime, and liquid splash.
- CPE is water resistant and provides good resistance against chemicals
- Economical and lint-free
Ideal for laboratory use.
* 通用尺寸
规格
- 滑轨尺寸:U*
- 颜色:蓝色
- 顺序:04
- Cat. No.:VWRU414004-512
- Garment application:Laboratory