Orders LinkContactUsLinkComponent
鞋套,CPE
鞋套,CPE
04, 蓝色, U*
目录编号 VWRU414004-512
鞋套,CPE
目录编号 VWRU414004-512
鞋套,CPE
鞋套,CPE
04, 蓝色, U*
Jump to:
Price
¥1,255.00
Log in 查看库存
1 * 1.000 件
Product Details & Documents
Basic shoe covers manufactured from a low-density chlorinated polyethylene fabric, which is designed to protect against dirt, grime, and liquid splash.
  • CPE is water resistant and provides good resistance against chemicals
  • Economical and lint-free
Ideal for laboratory use.
* 通用尺寸
规格
  • 滑轨尺寸:
    U*
  • 颜色:
    蓝色
  • 顺序:
    04
  • Cat. No.:
    VWRU414004-512
  • Garment application:
    Laboratory