Orders LinkContactUsLinkComponent
广口瓶,TraceClean®,琥珀色玻璃
广口瓶,TraceClean®,琥珀色玻璃
02, Not Processed (NPC), 250 ml, 固体盖
Catalog # VWRU89094-036
广口瓶,TraceClean®,琥珀色玻璃
Catalog # VWRU89094-036
广口瓶,TraceClean®,琥珀色玻璃
广口瓶,TraceClean®,琥珀色玻璃
02, Not Processed (NPC), 250 ml, 固体盖
Price
¥2,240.00
Log in for availability
1 * 24 件
Product Details & Documents
III型(碱石灰)琥珀色玻璃
广口瓶有全封闭式和开口式盖子可选。全封闭式盖子衬里由氟聚合物树脂制成,用于半挥发物、农药、PCB和酚类分析。开放型盖子隔膜由超声粘合的含氟聚合物树脂/硅酮制成,用于挥发性有机物分析。建议将琥珀色玻璃用于对紫外光敏感的样品。
处理(PC)和Quality-Assured® (QA)广口瓶,按照OSWER指令9240.0-05A“无污染物样品容器规范和指南”中规定的EPA清洁程序运行。每箱中有一份QA广口瓶分析证明书。QA广口瓶条形码带有批号和每个容器的编号,具有完全可追溯性。未处理(NPC)广口瓶不能接受专业清洗工艺。每箱均进行密封,确保箱内物品的完整性。
Specifications
  • 盖:
    固体盖
  • 容量:
    250 ml
  • 说明:
    Not Processed (NPC)
  • 顺序:
    02