Orders LinkContactUsLinkComponent
K型探头
K型探头
01, K-type fast response probe with 1,2 m coated cable, –40…+250 °C, 1,6×1200 mm
目录编号 VWRU61161-372
K型探头
目录编号 VWRU61161-372
K型探头
01, K-type fast response probe with 1,2 m coated cable, –40…+250 °C, 1,6×1200 mm
Jump to:
Price
¥980.00
Log in 查看库存
1 * 1 件
规格
  • 范围:
    –40…+250 °C
  • 说明:
    K-type fast response probe with 1,2 m coated cable
  • 顺序:
    01
  • 直径×长度:
    1,6×1200 mm