pH电极,pHenomenal®
002, pHenomenal® 111 pH电极,3合1,带温度传感器, pHenomenal® 111,3合1,凝胶*, 环氧树脂, 凝胶, 陶瓷, 12×120 mm, BNC,1 m电缆
目录编号 VWRI662-1157
供应商: VWR Collection
Jump to:
Product Details & Documents
pH electrodes with BNC connector and cable.
These electrodes are checked at five points during the manufacturing process and then lot stamped to ensure quality. Every electrode is supplied with a soaker storage bottle and cap to ensure the pH-sensitive glass and reference junction remain in top condition and are instantly ready for use.
* Integrated temperature probe.
Delivery information: Includes pH-electrode only. 620-1659 includes pHenomenal® TEMP21 T-sensor (NTC 30 kOhm) only.
Documents
CE Certificate_VWR Electrode.pdf CE_VWR_Phenomenal_pH_electrode_Conductivity_oxygen_meter.pdf EUDoC_portal_info
Manual VWR® Phenomenal pH Sensors Manual_VWR_IS_2000_L_Bedienungsanleitung_DE_270213 Manual_VWR_IS_2000_L_e_operativo_IT_270213 Manual_VWR_IS_2000_L_Instrucciones_de_operacion_ES_270213 Manual_VWR_IS_2000_L_Instrucoes_de_Utilizacao_PT_270213
Manual
More
VWR HSG Electrochemistry VWR pH Buffer Solutions VWR pH Electrodes
Product Literature
VWRI662-1161_TDS_EN
Datasheet
规格
- 连接:BNC,1 m电缆
- 直径×长度:12×120 mm
- Type:pHenomenal® 111,3合1,凝胶*
- 说明:pHenomenal® 111 pH电极,3合1,带温度传感器
- 材质:环氧树脂
- 顺序:002
- 隔膜:陶瓷
- 电解质:凝胶



