订单输入
China
Orders LinkContactUsLinkComponent
pH电极,pHenomenal®

pH电极,pHenomenal®

10, pHenomenal® TEMP21, 环氧树脂, 4 mm banana, m电缆
目录编号 VWRI620-1659
供应商:   VWR Collection
pH电极,pHenomenal®
目录编号 VWRI620-1659
供应商:  VWR Collection
pH电极,pHenomenal®
pH电极,pHenomenal®

10, pHenomenal® TEMP21, 环氧树脂, 4 mm banana, m电缆

Jump to:
Price
¥4,216.00
Log in 查看库存
1 * 1 件
Product Details & Documents

pH electrodes with BNC connector and cable.

These electrodes are checked at five points during the manufacturing process and then lot stamped to ensure quality. Every electrode is supplied with a soaker storage bottle and cap to ensure the pH-sensitive glass and reference junction remain in top condition and are instantly ready for use.

* Integrated temperature probe.

Delivery information: Includes pH-electrode only. 620-1659 includes pHenomenal® TEMP21 T-sensor (NTC 30 kOhm) only.

规格
  • 连接:
    4 mm banana, m电缆
  • Type:
    pHenomenal® TEMP21
  • 材质:
    环氧树脂
  • 顺序:
    10