订单输入
China
Orders LinkContactUsLinkComponent
大数字三显示温度计,Traceable

大数字三显示温度计,Traceable

Thermometer with waterproof bullet probe sensor, −50…+70 °C, ±1.0 °C, 0,1 °C, 防水bullet, 99×111×22 mm, 113 g
目录编号 VWRI620-1585
供应商:   VWR Collection
大数字三显示温度计,Traceable
目录编号 VWRI620-1585
供应商:  VWR Collection
大数字三显示温度计,Traceable
大数字三显示温度计,Traceable

Thermometer with waterproof bullet probe sensor, −50…+70 °C, ±1.0 °C, 0,1 °C, 防水bullet, 99×111×22 mm, 113 g

Jump to:
Price
¥2,100.00
Log in 查看库存
1 * 1 件
Product Details & Documents

These jumbo size display thermometers are suitable for monitoring freezers, water baths, heating blocks, incubators and refrigerators. A bottle insulates the sensor from transient temperature changes when the refrigerator door is opened. Solution is GRAS (Generally Recognised As Safe) by the FDA (Food and Drug Administration).

  • Triple display: Maximum/minimum and current temperatures
  • Out of range alarm programmable in 1,0 °C increments

Bottle probe in non toxic glycol solution or waterproof bullet probe (20 mm).

Bottle probe sensor: Temperature-buffered glycol bottle probe mimics solution. Bottle and solution insulate sensor from rapid changes, as an example, when a refrigerator door is opened. Solution mimics temperature of stored samples. Probe is sealed in a miniature bottle filled with nontoxic glycol. Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration).

Bullet probe sensor: Small sized plastic probe designed for use in air and liquids, the sensor and cable may be completely immersed.

Certifications: Traceability to NIST. Fulfils CDC and vaccine thermometer requirements. Compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.

Delivery information: Supplied with Traceable® certificate of calibration, slot for wall mounting, VELCRO® and magnetic strips to attach to any surface, AAA battery and 3 metre cable.

规格
  • 范围:
    −50…+70 °C
  • 分辨率:
    0,1 °C
  • 重量:
    113 g
  • 说明:
    Thermometer with waterproof bullet probe sensor
  • 宽度×深度×高度:
    99×111×22 mm
  • 准确度:
    ±1.0 °C
  • 探针:
    防水bullet