大数字三显示温度计,Traceable
Product Details & Documents
These jumbo size display thermometers are suitable for monitoring freezers, water baths, heating blocks, incubators and refrigerators. A bottle insulates the sensor from transient temperature changes when the refrigerator door is opened. Solution is GRAS (Generally Recognised As Safe) by the FDA (Food and Drug Administration).
- Triple display: Maximum/minimum and current temperatures
- Out of range alarm programmable in 1,0 °C increments
Bottle probe in non toxic glycol solution or waterproof bullet probe (20 mm).
Bottle probe sensor: Temperature-buffered glycol bottle probe mimics solution. Bottle and solution insulate sensor from rapid changes, as an example, when a refrigerator door is opened. Solution mimics temperature of stored samples. Probe is sealed in a miniature bottle filled with nontoxic glycol. Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration).
Bullet probe sensor: Small sized plastic probe designed for use in air and liquids, the sensor and cable may be completely immersed.
Certifications: Traceability to NIST. Fulfils CDC and vaccine thermometer requirements. Compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Delivery information: Supplied with Traceable® certificate of calibration, slot for wall mounting, VELCRO® and magnetic strips to attach to any surface, AAA battery and 3 metre cable.
规格
- 范围:−50…+70 °C
- 分辨率:0,1 °C
- 重量:113 g
- 说明:Thermometer with bottle probe sensor
- 宽度×深度×高度:99×111×22 mm
- 准确度:±1.0 °C
- 探针:瓶



