Colour coders for cryovials, CryoColorCode, Nunc™
07, 橙色
Catalog # NUNC355158
Supplier: Thermo Fisher Scientific
Colour coders for cryovials, CryoColorCode, Nunc™
Catalog # NUNC355158
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Colour coded inserts for the cap with inscription area.
Notch to remove tube or CryoColorCodes.
Documents
Specifications
- 颜色:橙色
- 顺序:07
- Cat. No.:NUNC355158
- 环境认证:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact