Colour coders for cryovials, CryoColorCode, Nunc™
04, 绿色
目录编号 NUNC355018
Colour coders for cryovials, CryoColorCode, Nunc™
目录编号 NUNC355018
Jump to:
Product Details & Documents
Colour coded inserts for the cap with inscription area.
Notch to remove tube or CryoColorCodes.
规格
- 颜色:绿色
- 顺序:04
- Cat. No.:NUNC355018
- 环境认证:ISO 14001 (Certificate Registration No.CERT-IT000330)
- Environmentally Preferable:Low Manufacturing Impact