订单输入
China
Orders LinkContactUsLinkComponent
Sample tubes

Sample tubes

50 ml, 30 mm, 115 mm, 黄色, non sterile
目录编号 SIMPC571-2
供应商:   SIMPORT SCIENTIFIC
Sample tubes
目录编号 SIMPC571-2
供应商:  SIMPORT SCIENTIFIC
Sample tubes
Sample tubes

50 ml, 30 mm, 115 mm, 黄色, non sterile

Jump to:
Price
询价
Log in 查看库存
1 * 500 件
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These tubes are chemically resistant and shatterproof, they are supplied with a leakproof screw cap, particularly important when transporting hazardous material.

  • Tube made of PP
  • Cap made of PE
  • Self-standing base with graduation
  • Leakproof screw cap
  • Available sterile and non-sterile
  • Functional temperature range: –90 to +121 °C

Conical bottom tubes with self-standing base graduated from 10 to 30 ml and 10 to 50 ml respectively in 5 ml increments. Ideal for transport and storage of urine, sputum and most liquids or particulate samples. Molded-in graduations make them easy to read.

Packaging: Please note that the non-sterile caps and tubes are package separately in packages of 100. Sterile tubes are supplied in packs of 25.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904

规格
  • 外径:
    30 mm
  • 版本:
    non sterile
  • 容量:
    50 ml
  • 高度:
    115 mm
  • 盖颜色:
    黄色

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.