Orders LinkContactUsLinkComponent
Test kits, digital titration method
16, Total hardness LR: EDTA method, 10 - 160 mg/l, 100 Tests
目录编号 HACH2448000
供应商:   HACH
Test kits, digital titration method
目录编号 HACH2448000
供应商:  HACH
CAS号:  
Test kits, digital titration method
Test kits, digital titration method
16, Total hardness LR: EDTA method, 10 - 160 mg/l, 100 Tests
Jump to:
CAS号:  
Price
询价
Log in 查看库存
1 * 100 Tests
Product Details & Documents
Digital titration provides a more precise and accurate method than the drop test method.
  • Reagents are pre-prepared
  • Exact comparison of colour
  • Rugged carrying case for analysis in the field
 : Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
规格
规格
  • 包装规格:
    100 Tests
  • 说明:
    Total hardness LR: EDTA method
  • 顺序:
    16
  • 范围:
    10 - 160 mg/l
规格