VWR®, Camera Adapters
05, C-mount adapter 1,0×
Catalog # VWRI630-0868
Supplier: VWR Collection
Jump to:
Product Details & Documents
These adapters enable cameras with different chip sizes to be connected to microscopes with a C-mount thread.
Specifications
- 说明:C-mount adapter 1,0×
- 顺序:05