VWR®, Camera Adapters
01, C-mount adapter 0,4×
目录编号 VWRI630-0867
供应商: VWR Collection
Jump to:
Product Details & Documents
These adapters enable cameras with different chip sizes to be connected to microscopes with a C-mount thread.
规格
- 说明:C-mount adapter 0,4×
- 顺序:01