VWR®, Camera Adapters
04, C-mount adapter 1,0×
目录编号 VWRI630-0866
供应商: VWR Collection
Jump to:
Product Details & Documents
These adapters enable cameras with different chip sizes to be connected to microscopes with a C-mount thread.
规格
- 说明:C-mount adapter 1,0×
- 顺序:04