校准重量,1级
07, 5000 g, 12000 ± mg
目录编号 VWRI611-8765
供应商: VWR Collection
Jump to:
Product Details & Documents
调整至 1级的不锈钢块,用于电子天平的精密检查和调整。
Certifications: ASTM certified.
Delivery information: Individual weights are supplied without certificate.
Documents
规格
- 重量:5000 g
- 顺序:07
- 误差:12000 ± mg