China
Orders LinkContactUsLinkComponent
校准重量,1级
校准重量,1级
04, 500 g, 1200 ± mg
目录编号 VWRI611-8762
供应商:   VWR Collection
校准重量,1级
目录编号 VWRI611-8762
供应商:  VWR Collection
校准重量,1级
校准重量,1级
04, 500 g, 1200 ± mg
Jump to:
Price
¥3,976.00
Log in 查看库存
1 * 1 件
Product Details & Documents
调整至 1级的不锈钢块,用于电子天平的精密检查和调整。
Certifications: ASTM certified.
Delivery information: Individual weights are supplied without certificate.
规格
  • 重量:
    500 g
  • 顺序:
    04
  • 误差:
    1200 ± mg