校准重量,1级
02, 100 g, 250 ± mg
Catalog # VWRI611-8760
Supplier: VWR Collection
Jump to:
Product Details & Documents
调整至 1级的不锈钢块,用于电子天平的精密检查和调整。
Certifications: ASTM certified.
Delivery information: Individual weights are supplied without certificate.
Documents
VWR® for Calibrations VWR® for Weighing
Product Literature
Specifications
- 重量:100 g
- 顺序:02
- 误差:250 ± mg