Tip refill systems, Axygen®
02, 1 - 200 µl, -, 透明, 10 racks, 96 each (stack rack)
目录编号 AXYGRFL-222-C
供应商: Corning
Jump to:
Product Details & Documents
Transparent PP, autoclavable.
- Easy to load refill racks
- Certified RNase-, DNase- and endotoxin-free
- Universal fit tips have a bevelled reference and graduated marks
Documents
规格
- 包装规格:10 racks, 96 each (stack rack)
- 灭菌:-
- 容积:1 - 200 µl
- 顺序:02
- 尖端颜色:透明