Filter funnels
09, 125 ml, 60 mm, 1
目录编号 LENZ07195140
供应商: Lenz Laborglas
Jump to:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filters disc.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
规格
- Plate Ø:60 mm
- 多孔性:1
- 容量:125 ml
- 顺序:09