Adapters, PTFE, socket on cone
08, full length, A, 29/42 NS, 24/40 NS
目录编号 COWI021.02429
供应商: COWIE
Jump to:
Product Details & Documents
Made of pure PTFE, white.
- Very good chemical resistance
- Very good temperature resistance
- Fully adjustable with replacement Viton® seal
- Contact materials are glass and PTFE only
This holder enables standard probes and thermometers to be assembled in standard taper laboratory glassware.
规格
- 圆锥体大小:29/42 NS
- 插座尺寸:24/40 NS
- Type:full length, A
- 顺序:08