Adapters, PTFE, socket on cone
07, full length, A, 34/45 NS, 19/38 NS
Catalog # COWI021.01934
Supplier: COWIE
Jump to:
Product Details & Documents
Made of pure PTFE, white.
- Very good chemical resistance
- Very good temperature resistance
- Fully adjustable with replacement Viton® seal
- Contact materials are glass and PTFE only
This holder enables standard probes and thermometers to be assembled in standard taper laboratory glassware.
Specifications
- 圆锥体大小:34/45 NS
- 插座尺寸:19/38 NS
- Type:full length, A
- 顺序:07