Orders LinkContactUsLinkComponent
Adapters, PTFE, socket on cone
05, full length, A, 24/40 NS, 19/38 NS
目录编号 COWI021.01924
供应商:   COWIE
Adapters, PTFE, socket on cone
目录编号 COWI021.01924
供应商:  COWIE
Adapters, PTFE, socket on cone
Adapters, PTFE, socket on cone
05, full length, A, 24/40 NS, 19/38 NS
Jump to:
Price
询价
Log in 查看库存
1 * 1 件
Product Details & Documents
Made of pure PTFE, white.
  • Very good chemical resistance
  • Very good temperature resistance
  • Fully adjustable with replacement Viton® seal
  • Contact materials are glass and PTFE only
This holder enables standard probes and thermometers to be assembled in standard taper laboratory glassware.
规格
  • 圆锥体大小:
    24/40 NS
  • 插座尺寸:
    19/38 NS
  • Type:
    full length, A
  • 顺序:
    05