Digital Solutions
Applications
Support
About
China
Orders Link
ContactUsLinkComponent
缓冲(液)
Cloning Buffers
Buffer, TE solution (TRIS-EDTA buffer)
Buffer, TE solution (TRIS-EDTA buffer)
TE buffer solution (TRIS-EDTA buffer), Glass bottle, 100 mL
PROMV6231
:
PROMEGA
Buffer, TE solution (TRIS-EDTA buffer)
PROMV6231
:
PROMEGA
:
Buffer, TE solution (TRIS-EDTA buffer)
TE buffer solution (TRIS-EDTA buffer), Glass bottle, 100 mL
:
1 * 100 毫升
-
+
包装类型:
Glass bottle
包装规格:
100 mL
说明:
TE buffer solution (TRIS-EDTA buffer)