Measuring scoops
100 ml, 200 mm
目录编号 VITL39694
供应商: VITLAB
Jump to:
Product Details & Documents
PP, white. With flat, chamfered ends and reinforced handle.
- Excellent chemical resistance, robust
- Heat resistant up to +130 °C, autoclavable
- The capacity of the scoop is stamped into the handle for easy reading
Documents
规格
- 容量:100 ml
- 长度:200 mm