Brinzolamide, Reference Standards
03, Brinzolamide Related Compound B, USP reference standard, Vial, 50 mg
Catalog # USPH1076385
Supplier: U.S. PHARMACOPEIA
Jump to:
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- 包装类型:Vial
- 包装规格:50 mg
- 标准:USP reference standard
- 说明:Brinzolamide Related Compound B
- 顺序:03
- Cat. No.:USPH1076385
- 公式:C₁₀H₁₇N₃O₅S₃ ,C₂H₂O₄
- 说明:Brinzolamide Related Compound B
- Synonym(s):(R)-4-Amino-2-(3-methoxypropyl)-3,4-dihydro-2H-thieno[3,2-e][1,2]thiazine-6-sulfonamide 1,1-dioxide oxalate
- CAS No.:404034-55-5



