Filter funnels
03, 50 ml, 35 mm, 3
Catalog # LENZ07195328
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
Borosilicate glass 3.3.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filters disc.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Plate Ø:35 mm
- 多孔性:3
- 容量:50 ml
- 顺序:03