Biopsy processing/embedding cassettes, Microsette™ M503
橘粉色
目录编号 SIMPM503-7
供应商: SIMPORT SCIENTIFIC
Jump to:
Product Details & Documents
Acetal polymer.
- No need for biopsy pads
- Six compartments, each 7×12 mm, numbered 1 to 6
- 2000 square openings (each 0,26 mm) to maximise fluid exchange
- Integral lid
- Anterior writing area 45° angle suitable for some labelling systems
规格
- 颜色:橘粉色