Orders LinkContactUsLinkComponent
Test kits, drop count tests
17, Total hardness, model 5-EP, 17 - 510 mg/l, 100, 1 KIT
目录编号 HACH145400
供应商:   HACH
Test kits, drop count tests
目录编号 HACH145400
供应商:  HACH
CAS号:  
Test kits, drop count tests
Test kits, drop count tests
17, Total hardness, model 5-EP, 17 - 510 mg/l, 100, 1 KIT
Jump to:
CAS号:  
Price
询价
Log in 查看库存
1 * 1 KIT
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
 : Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
规格
规格
  • 包装规格:
    1 KIT
  • 说明:
    Total hardness, model 5-EP
  • 顺序:
    17
  • 每套测试:
    100
  • 范围:
    17 - 510 mg/l
规格