订单输入
China
Orders LinkContactUsLinkComponent
三显示温度计,Traceable

三显示温度计,Traceable

01, –50…+70 °C, ±1.0 °C, 1,0 °C, 防水, 70×20×120 mm, 113 g
目录编号 VWRI620-2023
供应商:   VWR Collection
三显示温度计,Traceable
目录编号 VWRI620-2023
供应商:  VWR Collection
三显示温度计,Traceable
三显示温度计,Traceable

01, –50…+70 °C, ±1.0 °C, 1,0 °C, 防水, 70×20×120 mm, 113 g

Jump to:
Price
¥1,701.00
Log in 查看库存
1 * 1 件
Product Details & Documents

These digital thermometers are suitable for monitoring of temperatures in freezers, water baths, heating blocks, incubators and refrigerators. The bottle-enclosed, temperature-buffered sensor is insulated from transient temperature changes. Micro cable allows refrigerator doors to close on it.

  • Maximum/minimum monitors high and low readings overnight, at the weekend or for any time period
  • Programmable alarm sounds continuously when temperature rises above/falls below set points and can be turned off manually
  • Visual and audible signals continue even if temperature returns to non alarm range

Waterproof probe (Lר: 19×8 mm) or bottle-buffered sensor, with 3 metre cable.

Conforms to ISO 17025 and is traceable to NIST.

Patented unit fullfils all CDC thermometer and vaccine thermometer requirements.

Certifications: Compliance with ISO/IEC 17025, ANSI/NCSL Z541-1 and 9001. Traceability to NIST. Patented unit fulfils all CDC thermometer and vaccine thermometer requirements.

Delivery information: Supplied with Traceable® certificate of calibration, flip stand, wall bracket, VELCRO mounting strips and batteries.

规格
  • 范围:
    –50…+70 °C
  • 分辨率:
    1,0 °C
  • 重量:
    113 g
  • 顺序:
    01
  • 宽度×深度×高度:
    70×20×120 mm
  • 准确度:
    ±1.0 °C
  • 探针:
    防水