订单输入
China
Orders LinkContactUsLinkComponent
Buffer solutions for hardness testing

Buffer solutions for hardness testing

01, Hardness 1 buffer solution, pH=10,1, 500 ml, 1
目录编号 HACH42449
供应商:   HACH
Buffer solutions for hardness testing
目录编号 HACH42449
供应商:  HACH
CAS号:  
Buffer solutions for hardness testing
Buffer solutions for hardness testing

01, Hardness 1 buffer solution, pH=10,1, 500 ml, 1

Jump to:
CAS号:  
Price
询价
Log in 查看库存
1 * 1 件
Product Details & Documents

This buffer is used for hardness and chelant determination by titration.

Documents

Certificates
按批号搜索
规格

规格

  • 包装规格:
    1
  • 说明:
    Hardness 1 buffer solution, pH=10,1, 500 ml
  • 顺序:
    01

规格