Micro pipettes, ringcaps®
100 µl, 125 mm, Standard
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, with ring mark, disposable.
- Colour coded according to ISO for clear identification
- Conformity certified
ISO 9000, DIN ISO 7550, IVD 98/79 EC
Certifications: ISO 9000, DIN ISO 7550, IVD 98/79 EC
Documents
Specifications
- Type:Standard
- 容量:100 µl
- 长度:125 mm