Micro pipettes, ringcaps®
40 µl, 125 mm, Standard
HIRS9600140
DURAN®, borosilicate glass, with ring mark, disposable.
- Colour coded according to ISO for clear identification
- Conformity certified
ISO 9000, DIN ISO 7550, IVD 98/79 EC
: ISO 9000, DIN ISO 7550, IVD 98/79 EC
- Type:Standard
- 容量:40 µl
- 长度:125 mm