Micro pipettes, ringcaps®
25 µl, 125 mm, Standard
目录编号 HIRS9600125
供应商: HIRSCHMANN
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, with ring mark, disposable.
- Colour coded according to ISO for clear identification
- Conformity certified
ISO 9000, DIN ISO 7550, IVD 98/79 EC
Certifications: ISO 9000, DIN ISO 7550, IVD 98/79 EC
Documents
规格
- Type:Standard
- 容量:25 µl
- 长度:125 mm