Microtubes, with Thread
24, Micro tubes with writing space and graduation, neutral screw cap assembled, sterile, with skirted base, 2,0 ml, 1000/case
目录编号 SARS72.694.006
Marketsource
供应商: SARSTEDT
Product Details & Documents
PP, translucent.
- Safe screw cap with O-ring
- Sterile or non sterile
- Autoclavable
For transport, storage and sample processing in the laboratory.
Documents
规格
- 包装规格:1000/case
- 容量:2,0 ml
- 说明:Micro tubes with writing space and graduation, neutral screw cap assembled, sterile, with skirted base
- 顺序:24