VWR® Rectangular Cover Glasses
13, 24×50 mm, #1,5
目录编号 VWRI631-0147
供应商: VWR Collection
Jump to:
Product Details & Documents
Rectangular cover glasses made of pure white Hydrolytic Class 1 glass.
Packaging: In plastic hinged lid boxes.
规格
- 数量:#1,5
- 长度×宽度:24×50 mm
- 顺序:13