VWR® Rectangular Cover Glasses
11, 24×40 mm, #1
Catalog # VWRI631-0145
Supplier: VWR Collection
Jump to:
Product Details & Documents
Rectangular cover glasses made of pure white Hydrolytic Class 1 glass.
Packaging: In plastic hinged lid boxes.
Specifications
- 数量:#1
- 长度×宽度:24×40 mm
- 顺序:11