VWR® Rectangular Cover Glasses
09, 24×32 mm, #1
目录编号 VWRI631-0143
供应商: VWR Collection
Jump to:
Product Details & Documents
Rectangular cover glasses made of pure white Hydrolytic Class 1 glass.
Packaging: In plastic hinged lid boxes.
规格
- 数量:#1
- 长度×宽度:24×32 mm
- 顺序:09