VWR® Rectangular Cover Glasses
06, 22×40 mm, #1,5
目录编号 VWRI631-0136
供应商: VWR Collection
Jump to:
Product Details & Documents
Rectangular cover glasses made of pure white Hydrolytic Class 1 glass.
Packaging: In plastic hinged lid boxes.
规格
- 数量:#1,5
- 长度×宽度:22×40 mm
- 顺序:06