VWR® Rectangular Cover Glasses
01, 18×24 mm, #1
供应商: VWR Collection
Jump to:
Product Details & Documents
Rectangular cover glasses made of pure white Hydrolytic Class 1 glass.
: In plastic hinged lid boxes.
规格
- 数量:#1
- 长度×宽度:18×24 mm
- 顺序:01
Rectangular cover glasses made of pure white Hydrolytic Class 1 glass.
: In plastic hinged lid boxes.