Orders LinkContactUsLinkComponent
Scoops
2500 ml, 365 mm
  RSGA758.150
 :   RSG
Scoops
  RSGA758.150
 :  RSG
Scoops
Scoops
2500 ml, 365 mm
1 * 1 件
Stainless steel. Manual scoops for collecting or measuring powders, pastes or granulates.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Robust design
  • 容量:
    2500 ml
  • 长度:
    365 mm