Adapter, stopper type, DURAN®
stopper, solid, 29/32 NS, 24/29 NS
Catalog # SCOT286576906
Supplier: DWK LIFE SCIENCES
Jump to:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, stopper type, solid, socket and cone.
Specifications
- 圆锥体大小:29/32 NS
- 插座尺寸:24/29 NS
- Type:stopper, solid