pH indicator papers, MQuant®, Supelco®
Universal indicator, 1 - 14, 1
供应商: MERCK MILLIPORE
Jump to:
Product Details & Documents
Traditional pH indicator test papers, supplied on a 4,8 metre roll, consisting of high quality filter paper impregnated with indicator and mixed indicator solutions then dried and cut to size.
- Roll format are easy to use and most common ranges are available in a refill format
- Quality assurance - tests are checked and calibrated against certified buffer solutions
- Wide and narrow pH ranges available
* no colour scale card supplied
** pack of 3×100 strips
规格
- pH 等级:1
- pH 范围:1 - 14
- 说明:Universal indicator
- Cat. No.:MERC1.10962.0003
- M-Clarity quality level:MQ100