Spatulas, micro
Round end, 150 mm, 3 mm
Catalog # RSGA087.033
Supplier: RSG
Jump to:
Product Details & Documents
Stainless steel, 18/10. Micro spatulas with black plastic handle.
Specifications
- 说明:Round end
- 长度:150 mm
- 刀片宽度:3 mm